Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRTM2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298780MFV610
Description
LRRTM2 Polyclonal antibody specifically detects LRRTM2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
LRRTM2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
leucine-rich repeat transmembrane neuronal protein 2, leucine rich repeat transmembrane neuronal 2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human LRRTM2. | |
0.1 mL | |
Neuroscience | |
26045 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction