Learn More
Invitrogen™ LRRTM3 Polyclonal Antibody

Description
A suggested positive control is mouse brain tissue lysate. PA5-21098 can be used with blocking peptide PEP-1212.
Specifications
Specifications
| Antigen | LRRTM3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | LRRTM3 |
| Gene Accession No. | D3ZAL8, Q86VH5, Q8BZ81 |
| Gene Alias | 9630044H04Rik; leucine rich repeat transmembrane neuronal 3; leucine-rich repeat transmembrane neuronal protein 3; LRRTM3; UNQ803/PRO1693 |
| Gene Symbols | LRRTM3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.