Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRTM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113080
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113080 Supplier Invitrogen™ Supplier No. PA5113080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRTM4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Little is known about the function of LRRTM4; in mouse, it is expressed in the limb mesenchyme, neural tube, caudal mesoderm and in three distinct regions of the head. Later expression occurs in a subset of the developing sclerotome. Its similarity to other LRRTM family members suggests that LRRTM4 may also play a role in the development and maintenance of the vertebrate nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRTM4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LRRTM4
Gene Accession No. Q80XG9, Q86VH4
Gene Alias 7530419J18Rik; A230052N11; FLJ12568; leucine rich repeat transmembrane neuronal 4; leucine-rich repeat transmembrane neuronal 4 protein; leucine-rich repeat transmembrane neuronal protein 4; LRRTM4; MGC120633; MGC120636; RGD1560707; UNQ3075/PRO9907
Gene Symbols LRRTM4
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich repeat transmembrane neuronal protein 4 protein (259-419AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243499, 80059
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.