Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRSAM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP182016
Description
LRSAM1 Polyclonal specifically detects LRSAM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LRSAM1 | |
Polyclonal | |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
EC 6.3.2.-, FLJ31641, hTAL, leucine rich repeat and sterile alpha motif containing 1, Leucine-rich repeat and sterile alpha motif-containing protein 1, RIFLE, TALE3 ubiquitin-protein ligase LRSAM1, Tsg101-associated ligase | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
LRSAM1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:REILTELEAKSETRQENYWLIQYQRLLNQKPLSLKLQEEGMERQLVALLEELSAEHYLPIFAHHRLSLDLLSQMSPGDLAKVGVSEAGLQHEILR | |
0.1 mL | |
Lipid and Metabolism | |
90678 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction