Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRSAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540112
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540112 Supplier Invitrogen™ Supplier No. PA540112
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse spine core tissue. Predicted reactivity: Human (99%), Mouse (87%), Rat (88%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRSAM1
Applications Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% Thimerosal; pH 7
Gene Lrsam1
Gene Accession No. Q6UWE0, Q80ZI6
Gene Alias CMT2P; E3 ubiquitin-protein ligase LRSAM1; hTAL; leucine rich repeat and sterile alpha motif containing 1; leucine-rich repeat and sterile alpha motif-containing protein 1; LRSAM1; RGD1564403; RIFLE; RING-type E3 ubiquitin transferase LRSAM1; TAL; Tsg101-associated ligase; UNQ6496/PRO21356
Gene Symbols Lrsam1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LRSAM1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227738, 90678
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.