Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRSAM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576450
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding.
Specifications
LRSAM1 | |
Polyclonal | |
Unconjugated | |
Lrsam1 | |
CMT2P; E3 ubiquitin-protein ligase LRSAM1; hTAL; leucine rich repeat and sterile alpha motif containing 1; leucine-rich repeat and sterile alpha motif-containing protein 1; LRSAM1; RGD1564403; RIFLE; RING-type E3 ubiquitin transferase LRSAM1; TAL; Tsg101-associated ligase; UNQ6496/PRO21356 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
227738, 311866, 90678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWE0, Q80ZI6 | |
Lrsam1 | |
Recombinant fusion protein of human LRSAM1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction