Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598994
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598994 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598994 Supplier Invitrogen™ Supplier No. PA598994
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRTM1 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

LRTM1 is a protein coding gene and gene ontology (GO) annotation includes extracellular space; integral component of membrane; heparin binding; Roundabout binding; axon guidance; negative chemotaxis; positive regulation of synapse assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRTM1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LRTM1
Gene Accession No. Q9HBL6
Gene Alias HT017; leucine rich repeats and transmembrane domains 1; leucine-rich repeat and transmembrane domain-containing protein 1; leucine-rich repeats and transmembrane domains 1; LRTM1
Gene Symbols LRTM1
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich repeat and transmembrane domain-containing protein 1 protein (1-212AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57408
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.