Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRTM2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306292MFV610
Description
LRTM2 Polyclonal antibody specifically detects LRTM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
LRTM2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Leucine-Rich Repeat And Transmembrane Domain-Containing Protein 2, Leucine-Rich Repeats And Transmembrane Domains 2 | |
Produced in rabbits immunized with E. coli-derived Human LRTM2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
654429 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction