Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRTM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598733 Supplier Invitrogen™ Supplier No. PA598733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRTM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

LRTM2 is a protein coding gene and gene ontology (GO) includes cytoplasm, protein kinase inhibitor activity, cytokine-mediated signaling pathway, negative regulation of JAK-STAT cascade, negative regulation of protein kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRTM2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LRTM2
Gene Accession No. Q8N967
Gene Alias leucine rich repeats and transmembrane domains 2; leucine-rich repeat and transmembrane domain-containing protein 2; leucine-rich repeats and transmembrane domains 2; LRTM2
Gene Symbols LRTM2
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich repeat and transmembrane domain-containing protein 2 protein (36-310AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 654429, 680883
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.