Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRTM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563062
Description
Immunogen sequence: NNSIRTLDRD LLRHSPLLRH LDLSINGLAQ LPPGLFDGLL ALRSLSLRSN RLQNLDRLTF EPLANLQLLQ VGDNPWECDC NLREFKHWME Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.
LRTM2 is a protein coding gene and gene ontology (GO) includes cytoplasm, protein kinase inhibitor activity, cytokine-mediated signaling pathway, negative regulation of JAK-STAT cascade, negative regulation of protein kinase activity.
Specifications
LRTM2 | |
Polyclonal | |
Unconjugated | |
LRTM2 | |
leucine rich repeats and transmembrane domains 2; leucine-rich repeat and transmembrane domain-containing protein 2; leucine-rich repeats and transmembrane domains 2; LRTM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
654429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N967 | |
LRTM2 | |
Recombinant protein corresponding to Human LRTM2. Recombinant protein control fragment (Product #RP-101435). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction