Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRTOMT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559005
Description
Immunogen sequence: MNKRDYMNTS VQEPPLDYSF RSIHVIQDLV NEEPRTGLRP LKRSKSGKSL TQSLWLNNNV LNDLRDFNQV ASQLLEHPEN LAWIDLSFND LTSIDPV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
Catalyzes the O-methylation, and thereby the inactivation, of catecholamine neurotransmitters and catechol hormones. Required for auditory function.
Specifications
LRTOMT | |
Polyclonal | |
Unconjugated | |
LRTOMT | |
1700008D07Rik; Catechol O-methyltransferase 2; catechol O-methyltransferase 2 {ECO:0000250; CFAP111; Comt2; DFNB63; leucine rich repeat containing 51; leucine rich transmembrane and 0-methyltransferase domain containing; leucine rich transmembrane and O-methyltransferase domain containing; leucine-rich repeat-containing protein 51; leucine-rich repeat-containing protein 51 {ECO:0000312; leucine-rich repeat-containing protein 51; transmembrane O-methyltransferase; LRRC51; lrrc51 {ECO:0000312; Lrtomt; Lrtomt1; O-methyltransferase domain containing; PP7517; Protein LRTOMT1; protein LRTOMT2; RGD:1561509}; RGD:1565856}; RGD1561509; RGD1565856; Tomt; tomt {ECO:0000312; Transmembrane O-methyltransferase; transmembrane O-methyltransferase {ECO:0000312; Transmembrane O-methyltransferase homolog; UniProtKB:A1Y9I9} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220074 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WZ04 | |
LRTOMT | |
Recombinant protein corresponding to Human LRTOMT. Recombinant protein control fragment (Product #RP-97527). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction