Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRWD1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297339AF750
Description
LRWD1 Polyclonal specifically detects LRWD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
LRWD1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DKFZp434K1815, leucine-rich repeats and WD repeat domain containing 1, ORCAleucine-rich repeat and WD repeat-containing protein 1, ORC-associated protein, origin recognition complex associated, Origin recognition complex-associated protein | |
Produced in rabbits immunized with E. coli-derived Human LRWD1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
222229 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction