Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRWD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554380 Supplier Invitrogen™ Supplier No. PA554380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFLTVNDNLK VSFLLPTLRK VNGKDASSTY SQVENLNREL TSRVTAHWEK FMATLGPEEE AEKAQADFVK SAVRD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.

Required for G1/S transition. Recruits and stabilizes the origin recognition complex (ORC) onto chromatin during G1 to establish pre-replication complex (preRC) and to heterochromatic sites in post-replicated cells. Binds a combination of DNA and histone methylation repressive marks on heterochromatin. Binds histone H3 and H4 trimethylation marks H3K9me3, H3K27me3 and H4K20me3 in a cooperative manner with DNA methylation. Required for silencing of major satellite repeats. May be important ORC2, ORC3 and ORC4 stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRWD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRWD1
Gene Accession No. Q9UFC0
Gene Alias 1200011O22Rik; AU042569; AW548074; CENP-33; Centromere protein 33; leucine rich repeats and WD repeat domain containing 1; leucine-rich repeat and WD repeat-containing protein 1; leucine-rich repeats and WD repeat domain containing 1; LOC304396; Lrwd1; ORCA; ORC-associated protein; Origin recognition complex-associated protein; similar to hypothetical protein DKFZp434K1815; testicular tissue protein Li 4
Gene Symbols LRWD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRWD1. Recombinant protein control fragment (Product #RP-92783).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222229
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.