Learn More
LRWD1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301867AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Required for G1/S transition. Recruits and stabilizes the origin recognition complex (ORC) onto chromatin during G1 to establish pre-replication complex (preRC) and to heterochromatic sites in post-replicated cells. Binds a combination of DNA and histone methylation repressive marks on heterochromatin. Binds histone H3 and H4 trimethylation marks H3K9me3, H3K27me3 and H4K20me3 in a cooperative manner with DNA methylation. Required for silencing of major satellite repeats. May be important ORC2, ORC3 and ORC4 stability.Specifications
| LRWD1 | |
| Polyclonal | |
| Unconjugated | |
| LRWD1 | |
| CENP-33, ORCA, ORC-associated protein, centromere protein 33, leucine-rich repeat and WD repeat-containing protein 1, origin recognition complex associated, origin recognition complex-associated protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222229 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9UFC0 | |
| LRWD1 | |
| Between 200 and 250 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.