Learn More
Invitrogen™ LSAMP Polyclonal Antibody

Description
Immunogen sequence: VTNASLVLFRP GSVRGINGSI Highest antigen sequence identity to the following orthologs - mouse 95%, rat 100%.
Specifications
Specifications
| Antigen | LSAMP |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Lsamp |
| Gene Accession No. | Q13449 |
| Gene Alias | 5430428I19; AW046674; B130007O04Rik; D930023J12Rik; IgLON family member 3; IGLON3; Lam; LAMP; limbic system associated membrane protein; limbic system-associated membrane protein; Lsamp; neuronal growth regulator 1-like; Unknown (protein for MGC:128495) |
| Gene Symbols | Lsamp |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.