Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSD1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100536UV
Description
LSD1 Polyclonal antibody specifically detects LSD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
LSD1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human FAD-binding protein BRAF35-HDAC complex 110 kDa using the numbering given in TrEMBL entry O60341 (GeneID 23028). | |
0.1 mL | |
DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
23028 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction