Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSD1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056475C
Description
LSD1 Polyclonal specifically detects LSD1 in Human, Mouse, Rat, Canine, Primate, Zebrafish samples. It is validated for Western Blot.Specifications
LSD1 | |
Polyclonal | |
Western Blot | |
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KDM1A | |
A portion of amino acids 100-150 of human LSD1 was used as the immunogen. | |
0.1 mL | |
DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
Human, Mouse, Rat, Canine, Primate, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction