Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318684
This item is not returnable.
View return policy
Description
LSD1 Polyclonal antibody specifically detects LSD1 in Human samples. It is validated for Western Blot, ChIP assay, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay SequencingSpecifications
LSD1 | |
Polyclonal | |
Unconjugated | |
PBS | |
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
Polyclonal antibody raised in rabbit against human LSD1 (Lysine-specific demethylase 1), using a KLH-conjugated synthetic peptide from the inner part of the protein. | |
50 μg | |
DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
23028 | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2.2 mg/ml | |
Western Blot 1:4000, Chromatin Immunoprecipitation 1 ug/ChIP, ELISA 1:1000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200-1:500, Chromatin Immunoprecipitation Sequencing 1 ug/ChIP | |
O60341 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction