Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSD1/BHC110 Mouse anti-Human, Clone: AP06.20, MilliporeSigma™

Catalog No. 05939I100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05939I100UL 100 μL
05939I25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05939I100UL Supplier MilliporeSigma Supplier No. 05939I100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LSD1/BHC110 Monoclonal antibody specifically detects LSD1/BHC110 in Human samples. It is validated for ChIP assay (ChIP), Immunocytochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSD1/BHC110
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone AP06.20
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 116 amino acids from the N-terminal half of human Lysine-specific histone demethylase 1A (LSD1/BHC110).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.