Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSD1 Monoclonal Antibody (1B2E5)
GREENER_CHOICE

Catalog No. PIMA515273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515273 Supplier Invitrogen™ Supplier No. MA515273
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15273 targets LSD1/AOF2 in indirect ELISA, IHC and WB applications and shows reactivity with Human, mouse, and Non-human primate samples. The MA5-15273 immunogen is purified recombinant fragment of human LSD1 expressed in E. Coli. MA5-15273 detects LSD1/AOF2 which has a predicted molecular weight of approximately 93kDa.

LSD1 is a histone demethylase that specifically demethylates 'Lys-4' of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. LSD1 contains a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. It acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. LSD1 demethylates both mono- and tri-methylted 'Lys-4' of histone H3. This protein may play a role in the repression of neuronal genes. Alone, it is unable to demethylate H3 'Lys-4' on nucleosomes and requires the presence of RCOR1/CoREST to achieve such activity. It may also demethylate 'Lys-9' of histone H3, a specific tag for epigenetic transcriptional repression, thereby leading to derepression of androgen receptor target genes. Mutations affecting this gene can result in cleft palate, psychomotor retardation, and distintive facial features.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B2E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene KDM1A
Gene Accession No. O60341, Q6ZQ88
Gene Alias 1810043O07Rik; AA408884; amine oxidase (flavin containing) domain 2; AOF2; BHC110; BRAF35-HDAC complex protein BHC110; CPRF; D4Ertd478e; FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit; flavin-containing amine oxidase domain-containing protein 2; KDM1; KDM1A; KIAA0601; Lsd1; lysine (K)-specific demethylase 1; lysine (K)-specific demethylase 1A; lysine demethylase 1A; lysine-specific demethylase 1; lysine-specific histone demethylase 1; lysine-specific histone demethylase 1A; mKIAA0601; neuroprotective protein 3; RGD1562975; RP1-184J9.1; wu:fb82e06; wu:fc51a02; wu:fj36a05; wu:fj66b06
Gene Symbols KDM1A
Host Species Mouse
Immunogen Purified recombinant fragment of human LSD1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23028, 99982
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.