Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31864025UL
This item is not returnable.
View return policy
Description
LSD1 Polyclonal antibody specifically detects LSD1 in Human samples. It is validated for Western BlotSpecifications
LSD1 | |
Polyclonal | |
Western Blot 1:1000 | |
O60341 | |
Rabbit | |
Whole Antiserum | |
RUO | |
Primary | |
Human | |
Antisera |
Western Blot | |
Unconjugated | |
Whole antiserum | |
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A | |
Polyclonal antibody raised in rabbit against human LSD1 (Lysine-specific demethylase 1), using the full length recombinant protein. | |
25 μg | |
DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
23028 | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction