Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSDP5 (PLIN5) Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811701


Description
LSDP5 (PLIN5) Monoclonal antibody specifically detects LSDP5 (PLIN5) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
LSDP5 (PLIN5) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O75052 | |
Mouse | |
Protein A/G | |
RUO | |
9722 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3G4 | |
Unconjugated | |
NOS1AP | |
2310076L09Rik; AI415325; AW109675; lipid droplet associated protein; lipid droplet-associated protein PAT-1; Lipid storage droplet protein 5; LSDA5; LSDP5; Mldp; Myocardial LD protein; Oxpat; Pat1; PAT-1; perilipin 5; perilipin-5; Plin5 | |
Human recombinant protein fragment corresponding to amino acids 1-196 of human NOS1AP produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction