Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP217178
Description
LSG1 Polyclonal specifically detects LSG1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LSG1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
EC 3.6.1, EC 3.6.1.-, FLJ11301, FLJ27294, hLsg1, large subunit GTPase 1 homolog, large subunit GTPase 1 homolog (S. cerevisiae) | |
Rabbit | |
75 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9H089 | |
LSG1 | |
Recombinant protein encompassing a sequence within the center region of human LSG1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
55341 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction