Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546724 Supplier Invitrogen™ Supplier No. PA546724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VQAVMGYKPG SGVVTASTAS SENGAGKPWK KHGNRNKKEK SRRLYKHLDM Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes a protein related to the yeast large subunit GTPase 1. The encoded protein is necessary for cell viability and may localize in the endoplasmic reticulum, nucleus and cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSG1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LSG1
Gene Accession No. Q9H089
Gene Alias 5830465I20; AA409273; D16Bwg1547e; fb99b06; hLsg1; hypothetical protein LOC323464; large 60S subunit nuclear export GTPase 1; large subunit GTPase 1 homolog; large subunit GTPase 1 homolog (S. cerevisiae); LOW QUALITY PROTEIN: large subunit GTPase 1 homolog; LSG1; RGD1309089; Unknown (protein for MGC:133940); wu:fb99b06; zgc:76988
Gene Symbols LSG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LSG1 (aa 609-658).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55341
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.