Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LSG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558069
Description
Immunogen sequence: SELNDGYDWG RLNLQSVTEQ SSLDDFLATA ELAGTEFVAE KLNIKFVPAE ARTGLLSFEE SQRIKKLHEE NKQFLCIPRR PNWNQNTTPE E Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
This gene encodes a protein related to the yeast large subunit GTPase 1. The encoded protein is necessary for cell viability and may localize in the endoplasmic reticulum, nucleus and cytoplasm.
Specifications
| LSG1 | |
| Polyclonal | |
| Unconjugated | |
| LSG1 | |
| 5830465I20; AA409273; D16Bwg1547e; fb99b06; hLsg1; hypothetical protein LOC323464; large 60S subunit nuclear export GTPase 1; large subunit GTPase 1 homolog; large subunit GTPase 1 homolog (S. cerevisiae); LOW QUALITY PROTEIN: large subunit GTPase 1 homolog; LSG1; RGD1309089; Unknown (protein for MGC:133940); wu:fb99b06; zgc:76988 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 224092, 288029, 55341 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.40 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3UM18, Q5BJT6, Q9H089 | |
| LSG1 | |
| Recombinant protein corresponding to Human LSG1. Recombinant protein control fragment (Product #RP-96261). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction