Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSM1 Antibody (022), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290268JF646
Description
LSM1 Monoclonal antibody specifically detects LSM1 in Human samples. It is validated for Western Blot, ELISASpecifications
LSM1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
022 | |
Western Blot, ELISA | |
Cancer-associated Sm-like, CASMYJL124C, LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae), Small nuclear ribonuclear CaSm, U6 snRNA-associated Sm-like protein LSm1 | |
Rabbit immunized with purified, recombinant Human LSM1 (Uniprot#: O15116, Met1-Tyr133) | |
0.1 mL | |
Cancer | |
27257 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction