Learn More
Invitrogen™ LSM1 Monoclonal Antibody (4F7)

Description
Peptide Sequence: SIDQFANLVL HQTVERIHVG KKYGDIPRGI FVVRGENVVL LGEIDLEKES DTPLQQVSIE EILEEQRVEQ QTKLEAEKLK VQALKDRGLS IPRADTLDEY.
Specifications
Specifications
| Antigen | LSM1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4F7 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | LSM1 |
| Gene Accession No. | O15116 |
| Gene Alias | 2810025O06Rik; cancer-associated Sm protein; cancer-associated Sm protein; Cancer-associated Sm-like; cancer-associated Sm-like protein; CASM; Lsm1; LSM1 homolog, mRNA degradation associated; LSM1 homolog, U6 small nuclear RNA associated; LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM1 mRNA degradation associated; LSM1, U6 small nuclear RNA associated; LSM1-like protein U6 small nuclear RNA associated; small nuclear ribonuclear CaSm; U6 snRNA-associated Sm-like protein LSm1; YJL124C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.