Learn More
LSM1 Mouse anti-Human, Clone: OTI5E6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503121

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.Specifications
LSM1 | |
Monoclonal | |
Unconjugated | |
LSM1 | |
CASM, YJL124C | |
Mouse | |
Affinity Chromatography | |
RUO | |
27257 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15116 | |
LSM1 | |
Human recombit protein fragment corresponding to amino acids 1-133 of human LSM1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.