Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586751
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586751 Supplier Invitrogen™ Supplier No. PA586751
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene LSM1
Gene Accession No. O15116
Gene Alias 2810025O06Rik; cancer-associated Sm protein; cancer-associated Sm protein; Cancer-associated Sm-like; cancer-associated Sm-like protein; CASM; Lsm1; LSM1 homolog, mRNA degradation associated; LSM1 homolog, U6 small nuclear RNA associated; LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM1 mRNA degradation associated; LSM1, U6 small nuclear RNA associated; LSM1-like protein U6 small nuclear RNA associated; small nuclear ribonuclear CaSm; U6 snRNA-associated Sm-like protein LSm1; YJL124C
Gene Symbols LSM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in Human LSM1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.