Learn More
Invitrogen™ LSM1 Recombinant Rabbit Monoclonal Antibody (022)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529400
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human LSM1.
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifications
LSM1 | |
Recombinant Monoclonal | |
5 mg/mL | |
PBS with no preservative | |
O15116 | |
LSM1 | |
Recombinant Human LSM1 Protein (Met1-Tyr133). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
022 | |
Unconjugated | |
LSM1 | |
2810025O06Rik; cancer-associated Sm protein; cancer-associated Sm protein; Cancer-associated Sm-like; cancer-associated Sm-like protein; CASM; Lsm1; LSM1 homolog, mRNA degradation associated; LSM1 homolog, U6 small nuclear RNA associated; LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM1 mRNA degradation associated; LSM1, U6 small nuclear RNA associated; LSM1-like protein U6 small nuclear RNA associated; small nuclear ribonuclear CaSm; U6 snRNA-associated Sm-like protein LSm1; YJL124C | |
Rabbit | |
Protein A | |
RUO | |
27257 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.