Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Lsm10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569109
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
LSM10 gene ontology annotations related to this gene include positive regulation of G1/S transition of mitotic cell cycle.
Specifications
Lsm10 | |
Polyclonal | |
Unconjugated | |
LSM10 | |
Lsm10; LSM10, U7 small nuclear RNA associated; MST074; MSTP074; U7 snRNA-associated Sm-like protein LSm10; U7 snRNP-specific Sm-like protein LSM10 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
366468 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
LSM10 | |
synthetic peptide directed towards the N-terminal region of rat Lsm10. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction