Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lsm10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595808 Supplier Invitrogen™ Supplier No. PA595808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVSHSVKER TISENSLIIL LQGLQGRVTT VDLRDESVAH GRIDNVDAFM NIRLAKVTYT DRWGHQVKLD DLFVTGRNVR YVHIPDDVNI TSTIEQQLQI IHRVRNFGGK GQGRWEFPPK NCK; Positive Samples: Rat spleen, Rat lung, Rat testis; Cellular Location: Nucleus.

LSM10 gene ontology annotations related to this gene include positive regulation of G1/S transition of mitotic cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lsm10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LSM10
Gene Alias Lsm10; LSM10, U7 small nuclear RNA associated; MST074; MSTP074; U7 snRNA-associated Sm-like protein LSm10; U7 snRNP-specific Sm-like protein LSM10
Gene Symbols LSM10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-123 of human LSM10 (NP_1162701).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366468
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.