Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597938 Supplier Invitrogen™ Supplier No. PA597938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LSM11 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

LSm11 is a 360 amino acid protein that localizes to the nucleus and belongs to the LSm subfamily of snRNP Sm proteins. Containing an N-terminal domain that is necessary for pre-mRNA cleavage, LSm11 functions to binds specifically to U7 snRNA and is thought to play a role in cell cycle regulation and cell growth. The gene encoding LSm11 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM11
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LSM11
Gene Accession No. P83369
Gene Alias 2210404M20Rik; AV079530; FLJ38273; Lsm11; LSM11, U7 small nuclear RNA associated; RGD1565523; U7 snRNA-associated Sm-like protein LSm11; U7 snRNP-specific Sm-like protein LSM11
Gene Symbols LSM11
Host Species Rabbit
Immunogen Recombinant Human U7 snRNA-associated Sm-like protein LSm11 protein (1-141AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134353
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.