Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120408 Supplier Invitrogen™ Supplier No. PA5120408
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: LO2, Jurkat, HeLa, Mouse brain. Immunogen sequence: MAAPPGEYFS VGSQVSCRTC QEQRLQGEVV AFDYQSKMLA LKCPSSSGKP NHADILLINL QYVSEVEIIN DRTETPPPLA SLNVSKLASK ARTEKEEKLS QAYAISAGVS LEGQQLFQTI HKTIKDCKWQ EKNIVVMEEV VITPPYQVEN CKGKEGSALS HVRKIVEKHF RDVESQKILQ RSQAQQPQKE AALSS.

LSM12 gene ontology annotations related to this gene include RNA metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LSM12
Gene Accession No. Q3MHD2, Q9D0R8
Gene Alias 1110032E16Rik; 1110059P07; 2600001B17Rik; Lsm12; LSM12 homolog; LSM12 homolog (S. cerevisiae); PNAS-135; protein LSM12 homolog
Gene Symbols LSM12
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124801, 268490
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.