Learn More
LSM12 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304633A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
LSM12 gene ontology annotations related to this gene include RNA metabolic process.Specifications
LSM12 | |
Polyclonal | |
Unconjugated | |
LSM12 | |
Protein LSM12 homolog, LSM12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124801, 268490 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q3MHD2, Q9D0R8 | |
Lsm12 | |
Between 145 and 195 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.