Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSM14A Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299157AF350
Description
LSM14A Polyclonal specifically detects LSM14A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LSM14A | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
26065 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
alphaSNBP, C19orf13, DKFZp434D1335, FAM61A, family with sequence similarity 61, member A, hRAP55, hRAP55A, LSM14 homolog A, LSM14A, SCD6 homolog A (S. cerevisiae), Protein FAM61A, protein LSM14 homolog A, Protein SCD6 homolog, Putative alpha-synuclein-binding protein, RAP55ALSM14 homolog A (SCD6, S. cerevisiae), RAP55chromosome 19 open reading frame 13, RNA-associated protein 55, RNA-associated protein 55A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human LSM14A. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction