Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578464 Supplier Invitrogen™ Supplier No. PA578464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A549, H1299, HCT116, NT2D1 Predicted Reactivity: Pig (93%), Rhesus Monkey (97%), Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

LSM14A, Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM14A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Lsm14a
Gene Accession No. Q8K2F8, Q8ND56
Gene Alias 2700023B17Rik; AA407828; AlphaSNBP; AU017544; C19orf13; FAM61A; family with sequence similarity 61, member A; hRAP55; hRAP55A; LSM14 homolog A; LSM14 homolog A (SCD6, S. cerevisiae); LSM14A; LSM14A mRNA processing body assembly factor; LSM14A, mRNA processing body assembly factor; LSM14A, SCD6 homolog A; LSM14A, SCD6 homolog A (S. cerevisiae); mRAP55A; Protein FAM61A; Protein LSM14 homolog A; protein SCD6 homolog; Putative alpha-synuclein-binding protein; RAP55; RAP55A; RGD1305695; RNA-associated protein 55; RNA-associated protein 55A; Tral
Gene Symbols Lsm14a
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LSM14A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26065, 361554, 67070
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.