Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSM14A Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548713
Description
Predicted to react with bovine and mouse based on sequence homology.
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifications
LSM14A | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8ND56 | |
LSM14A | |
KLH conjugated synthetic peptide between 311-340 amino acids from the C-terminal region of human LSM14A. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
LSM14A | |
Protein LSM14 homolog A, Protein FAM61A, Protein SCD6 homolog, Putative alpha-synuclein-binding protein, AlphaSNBP, RNA-associated protein 55A, hRAP55, hRAP55A, LSM14A, C19orf13, FAM61A, RAP55, RAP55A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26065 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction