Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSM14B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318490
This item is not returnable.
View return policy
Description
LSM14B Polyclonal antibody specifically detects LSM14B in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
LSM14B | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
149986 | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot, Immunoprecipitation | |
1 mg/ml | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2-10 ug/mg lysate | |
bA11M20.3, C20orf40, FAM61B, FT005, LSM13, LSM14B LSM14B, SCD6 homolog B (S. cerevisiae), RAP55B | |
Recognizes region between residues Between between 335 and 385.. | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction