Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSM2 Antibody (1G7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281280R
Description
LSM2 Monoclonal specifically detects LSM2 in Human samples. It is validated for Western Blot, ELISA.Specifications
LSM2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
LSM2 | |
Recombinant human LSm2 (1-95aa) purified from E. coli | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
57819 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
1G7 | |
Western Blot, ELISA | |
C6orf28chromosome 6 open reading frame 28, G7b, LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae), Protein G7b, Small nuclear ribonuclear protein D homolog, snRNP, snRNP core Sm-like protein Sm-x5, U6 snRNA-associated Sm-like protein LSm2, YBL026W | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction