Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | LSM2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15750420
![]() |
Novus Biologicals
NBP15750420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157504
![]() |
Novus Biologicals
NBP157504 |
100 μL |
Each for $487.50
|
|
|||||
Description
LSM2 Polyclonal specifically detects LSM2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LSM2 | |
Polyclonal | |
Purified | |
RUO | |
Q9Y333 | |
57819 | |
Synthetic peptides corresponding to LSM2 (LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae)) The peptide sequence was selected from the middle region of LSM2. Peptide sequence TDPEKYPHMLSVKNCFIRGSVVRYVQLPADEVDTQLLQDAARKEALQQKQ | |
Primary | |
10 kDa |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
C6orf28chromosome 6 open reading frame 28, G7b, LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae), Protein G7b, Small nuclear ribonuclear protein D homolog, snRNP, snRNP core Sm-like protein Sm-x5, U6 snRNA-associated Sm-like protein LSm2, YBL026W | |
LSM2 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title