Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104191 Supplier Invitrogen™ Supplier No. PA5104191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LSM2.

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LSM2
Gene Accession No. O35900, Q9Y333
Gene Alias C6orf28; D17H6S56E2; D17H6S56E-2; Dmapl; Dmpkap; dystrophia myotonica-protein kinase associated protein; G7B; G7b alternative form; LSM2; LSM2 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM2 homolog, U6 small nuclear RNA associated; LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM2 U6 small nuclear RNA and mRNA degradation associated; protein G7b; small nuclear ribonuclear protein D homolog; small ribonuclear protein G7b; SmX5; Sm-X5; snRNP; snRNP core protein SMX5; snRNP core Sm-like protein Sm-x5; U6 snRNA-associated Sm-like protein LSm2; YBL026W
Gene Symbols LSM2
Host Species Rabbit
Immunogen A synthesized peptide derived from human LSM2(Accession Q9Y333), corresponding to amino acid residues H30-Q80.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27756, 57819, 684148
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.