Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LSM3 Monoclonal Antibody (01)
LSM3 Monoclonal Antibody (1)
Supplier: Thermo Scientific MA530653
Description

Specifications
LSM3 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P62310 | |
LSM3 | |
Recombinant Human LSM3 Protein (Met1-Gly102). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
01 | |
Unconjugated | |
LSM3 | |
1010001J12Rik; 2610005D18Rik; 6030401D18Rik; Lsm3; LSM3 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM3 homolog, U6 small nuclear RNA associated; LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM3 U6 small nuclear RNA and mRNA degradation associated; MDS017; SMX4; U6 snRNA-associated Sm-like protein LSm3; USS2; YLR438C | |
Mouse | |
Protein A | |
RUO | |
27258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction