Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LSM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560875
Description
Immunogen sequence: DDVDQQQTTN TVEEPLDLIR LSLDERIYVK MRNDRELRGR LHAYDQHLNM ILGDVEETVT TIEIDEETYE EIYKSTKRNI PMLFVRGDG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifications
LSM3 | |
Polyclonal | |
Unconjugated | |
LSM3 | |
1010001J12Rik; 2610005D18Rik; 6030401D18Rik; Lsm3; LSM3 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM3 homolog, U6 small nuclear RNA associated; LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM3 U6 small nuclear RNA and mRNA degradation associated; MDS017; SMX4; U6 snRNA-associated Sm-like protein LSm3; USS2; YLR438C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P62310 | |
LSM3 | |
Recombinant protein corresponding to Human LSM3. Recombinant protein control fragment (Product #RP-96453). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction