Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559754 Supplier Invitrogen™ Supplier No. PA559754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSGILKGFDP LLNLVLDGTI EYMRDPDDQY KLTEDTRQLG LVVCRGTSVV LICPQDGMEA IPNPFIQQQD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LSM7
Gene Accession No. Q9UK45
Gene Alias 0910001B06Rik; 1110033F18Rik; LSM7; LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM7 homolog, U6 small nuclear RNA associated; LSM7 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM7 U6 small nuclear RNA and mRNA degradation associated; U6 snRNA-associated Sm-like protein LSm7; YNL147W
Gene Symbols LSM7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LSM7. Recombinant protein control fragment (Product #RP-99085).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51690
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.