Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143997 Supplier Invitrogen™ Supplier No. PA5143997
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HL-60 whole cell, human HEPG2 whole cell, human U937 whole cell, human HEK293 whole cell, human MCF-7 whole cell, human Jurkat whole cell, human Raji whole cell, rat brain tissue, mouse brain tissue. IHC: human bladder cancer tissue, mouse brain tissue, rat brain tissue, mouse brain tissue, rat brain tissue, rat cerebellum tissue, human melanoma tissue, human ovary cancer tissue, human appendicitis tissue, human bladder cancer tissue, human gastric cancer tissue, human renal carcinoma tissue, human skin cancer tissue, human tonsil tissue, mouse cerebellum tissue. ICC/IF: MCF-7 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM7
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene LSM7
Gene Accession No. Q9CQQ8, Q9UK45
Gene Alias 0910001B06Rik; 1110033F18Rik; LSM7; LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM7 homolog, U6 small nuclear RNA associated; LSM7 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM7 U6 small nuclear RNA and mRNA degradation associated; U6 snRNA-associated Sm-like protein LSm7; YNL147W
Gene Symbols LSM7
Host Species Rabbit
Immunogen E.coli-derived human LSM7 recombinant protein (Position: M1-A103).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362829, 51690, 66094
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.