Learn More
LSM7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305099A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Binds specifically to the 3′-terminal U-tract of U6 snRNA and is probably a component of the spliceosome.Specifications
LSM7 | |
Polyclonal | |
Unconjugated | |
LSM7 | |
U6 snRNA-associated Sm-like protein LSm7, LSM7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51690 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9UK45 | |
LSM7 | |
Between 53 and 103 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.