Learn More
Invitrogen™ LSM8 Monoclonal Antibody (6B11)

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HL-60 whole cell, human Jurkat whole cell, human THP-1 whole, human K562 whole cell, rat testis tissue, rat pancrease tissue, mouse testis tissue, mouse pancrease tissue. IHC: human renal clear cell carcinoma tissue, human colonic adenocarcinoma tissue, human placenta tissue, human spleen tissue. ICC/IF: Hela cell. Flow: JK cell, RH35 cell, RAW2647 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | LSM8 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6B11 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | LSM8 |
| Gene Accession No. | O95777, Q6ZWM4 |
| Gene Alias | 2010003I05Rik; AW214405; Lsm8; LSM8 homolog, U6 small nuclear RNA associated; LSM8 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM8 U6 small nuclear RNA associated; MAK31-like protein; Naa38; N-alpha-acetyltransferase 38, NatC auxiliary subunit; N-alpha-acetyltransferase 38, NatC auxiliary subunit; LSM8 homolog, U6 small nuclear RNA associated; U6 snRNA-associated Sm-like protein LSm8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.