Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554154 Supplier Invitrogen™ Supplier No. PA554154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALENYINRTV AVITSDGRMI VGTLKGFDQT INLILDESHE RVFSSSQGVE QVVLGLYIVR GDNVAVIGEI DEETDSALDL GNIRAEP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a member of the like-Sm family of proteins. The encoded protein consists of a closed barrel shape, made up of five anti-parallel beta strands and an alpha helix. This protein partners with six paralogs to form a heteroheptameric ring which transiently binds U6 small nuclear RNAs and is involved in the general maturation of RNA in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LSM8
Gene Accession No. O95777
Gene Alias 2010003I05Rik; AW214405; Lsm8; LSM8 homolog, U6 small nuclear RNA associated; LSM8 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM8 U6 small nuclear RNA associated; MAK31-like protein; Naa38; N-alpha-acetyltransferase 38, NatC auxiliary subunit; N-alpha-acetyltransferase 38, NatC auxiliary subunit; LSM8 homolog, U6 small nuclear RNA associated; U6 snRNA-associated Sm-like protein LSm8
Gene Symbols LSM8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LSM8. Recombinant protein control fragment (Product #RP-92711).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.