Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LSM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144015
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144015 Supplier Invitrogen™ Supplier No. PA5144015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HELA whole cell, human HL-60 whole cell, human HEPG2 whole cell, human U937 whole cell, human Jurkat whole cell, human Raji whole cell, human THP-1 whole cell, human K562 whole cell, rat testis tissue, rat lung tissue, rat liver tissue, rat pancreas tissue, mouse testis tissue, mouse lung tissue, mouse liver tissue, mouse HEPA1-6 whole cell. IHC: human skin cancer tissue, mouse testis tissue, rat testis tissue, human ovary cancer tissue, human appendicitis tissue, human renal carcinoma tissue, human gastric cancer tissue, human melanoma tissue, human bladder cancer tissue, human tonsil tissue. ICC/IF: A431 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a member of the like-Sm family of proteins. The encoded protein consists of a closed barrel shape, made up of five anti-parallel beta strands and an alpha helix. This protein partners with six paralogs to form a heteroheptameric ring which transiently binds U6 small nuclear RNAs and is involved in the general maturation of RNA in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM8
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene LSM8
Gene Accession No. O95777, Q6ZWM4
Gene Alias 2010003I05Rik; AW214405; Lsm8; LSM8 homolog, U6 small nuclear RNA associated; LSM8 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM8 U6 small nuclear RNA associated; MAK31-like protein; Naa38; N-alpha-acetyltransferase 38, NatC auxiliary subunit; N-alpha-acetyltransferase 38, NatC auxiliary subunit; LSM8 homolog, U6 small nuclear RNA associated; U6 snRNA-associated Sm-like protein LSm8
Gene Symbols LSM8
Host Species Rabbit
Immunogen E.coli-derived human LSM8 recombinant protein (Position: M1-H96).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296913, 51691, 76522
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.